Assay Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ311771.1 | - | 7 - 8 | 992 | 60 | |
| AJ311772.1 | - | 5 - 6 | 900 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ311771.1 | - | 7 - 8 | 992 | 60 | |
| AJ311772.1 | - | 5 - 6 | 900 | 60 | ||
Set Membership: |
|