Assay Details
| Gene Symbol: Otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290536.1 | NP_001277465.1 | 3 - 4 | 1041 | 83 | |
| NM_001290537.1 | NP_001277466.1 | 4 - 5 | 661 | 83 | ||
| NM_138604.3 | NP_613070.2 | 3 - 4 | 1041 | 83 | ||
| GenBank mRNA | AF229642.1 | - | 3 - 4 | 182 | 83 | |
| AK010750.1 | - | 4 - 5 | 659 | 83 | ||
| AK149777.1 | - | 4 - 5 | 662 | 83 | ||
| AK155131.1 | - | 3 - 4 | 1017 | 83 | ||
| AK169921.1 | - | 5 - 6 | 627 | 83 | ||
| AK171874.1 | - | 3 - 4 | 782 | 83 | ||
| AK204980.1 | - | 2 - 3 | 134 | 83 | ||
| BC016529.1 | - | 3 - 4 | 1041 | 3594583 | 83 | |
| BC051111.1 | - | 3 - 4 | 780 | 5253679 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud5
Genomic Map