Assay Details
| Gene Symbol: Otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290536.1 | NP_001277465.1 | 4 - 5 | 1200 | 90 | |
| NM_001290537.1 | NP_001277466.1 | 5 - 6 | 820 | 90 | ||
| NM_138604.3 | NP_613070.2 | 4 - 5 | 1200 | 90 | ||
| GenBank mRNA | AK010750.1 | - | 5 - 6 | 818 | 90 | |
| AK149777.1 | - | 5 - 6 | 821 | 90 | ||
| AK167226.1 | - | 3 - 4 | 513 | 90 | ||
| AK169921.1 | - | 6 - 7 | 786 | 90 | ||
| AK171874.1 | - | 4 - 5 | 941 | 90 | ||
| BC016529.1 | - | 4 - 5 | 1200 | 3594583 | 90 | |
| BC051111.1 | - | 4 - 5 | 939 | 5253679 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud5
Genomic Map