Assay Details
| Gene Symbol: Otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290536.1 | NP_001277465.1 | 6 - 7 | 1547 | 74 | |
| NM_001290537.1 | NP_001277466.1 | 7 - 8 | 1167 | 74 | ||
| NM_138604.3 | NP_613070.2 | 6 - 7 | 1547 | 74 | ||
| GenBank mRNA | AF229642.1 | - | 6 - 7 | 688 | 74 | |
| AK009620.1 | - | 1 - 2 | 63 | 74 | ||
| AK010750.1 | - | 7 - 8 | 1167 | 74 | ||
| AK149777.1 | - | 7 - 8 | 1168 | 74 | ||
| AK155131.1 | - | 6 - 7 | 1523 | 74 | ||
| AK167226.1 | - | 5 - 6 | 860 | 74 | ||
| AK171874.1 | - | 6 - 7 | 1288 | 74 | ||
| BC016529.1 | - | 6 - 7 | 1547 | 3594583 | 74 | |
| BC051111.1 | - | 6 - 7 | 1286 | 5253679 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud5
Genomic Map