Assay Details
| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012266.1 | - | 3 - 4 | 498 | 98 | |
| AK027977.1 | - | 3 - 4 | 470 | 98 | ||
| BC004620.1 | - | 3 - 4 | 476 | 3585323 | 98 | |
| BC004703.1 | - | 3 - 4 | 1108 | 3499265 | 98 | |
| DV661455.1 | - | 2 - 3 | 72 | 98 | ||
| X97755.1 | - | 3 - 4 | 474 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ebp
Genomic Map