Assay Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009593.2 | NP_033723.1 | 2 - 3 | 406 | 109 | |
| GenBank mRNA | AF323659.1 | - | 2 - 3 | 402 | 109 | |
| AK037433.1 | - | 2 - 3 | 326 | 109 | ||
| BC119471.2 | - | 2 - 3 | 354 | 40048166 | 109 | |
| U34920.1 | - | 2 - 3 | 364 | 109 | ||
| Z48745.1 | - | 2 - 3 | 393 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcg1
Genomic Map