| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700004914.mm.1 |
NM_001163748.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004914.mm.1 |
NM_001163748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004914.mm.1 |
NM_008804.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004914.mm.1 |
NM_008804.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004914.mm.1 |
TC1700000535.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004916.mm.1 |
NM_001163748.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004916.mm.1 |
NM_001163748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004916.mm.1 |
NM_008804.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004916.mm.1 |
NM_008804.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004916.mm.1 |
TC1700000535.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004915.mm.1 |
NM_001163748.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004915.mm.1 |
NM_001163748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004915.mm.1 |
NM_008804.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004915.mm.1 |
NM_008804.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004915.mm.1 |
TC1700000535.mm.1 |
MTA TC |