Assay Details
| Gene Symbol: Tmem158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002267.2 | NP_001002267.2 | 1 | 1494 | 135 | |
| GenBank mRNA | AK009669.1 | - | 1 | 1469 | 135 | |
| BE631438.1 | - | 1 | 135 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894027 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894035 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894025 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894026 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967227 | NM_001002267.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967227 | AB114272.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967227 | AK009669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967227 | BC139427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967227 | BC139428.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125427 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141034 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967227 | NM_001002267.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967227 | AB114272.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967227 | AK009669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967227 | BC139427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967227 | BC139428.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26894050 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125427 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141034 | - | - |
| Whole Mouse Genome G4122A | A_51_P104012 | BE631438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104012 | NM_001002267.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104012 | AK009669.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101280 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141034 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125427 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07955129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08172461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07955120 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08172462 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894035 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894026 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894029 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894025 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26894027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26894003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07652289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07990527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07900421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07997694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07585656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07997707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07990544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07933366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07990542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07918184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07669474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26894055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10597946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597947 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437750_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452652_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428074_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597945 | - | - |
| Mouse Genome 430 2.0 Array | 1437750_at | - | - |
| Mouse Genome 430 2.0 Array | 1452652_at | - | - |
| Mouse Genome 430 2.0 Array | 1437750_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428074_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452652_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428074_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72309_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72309_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900027840.mm.1 | NM_001002267.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027840.mm.1 | NM_001002267.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027840.mm.1 | TC0900003301.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428074_at | - | - |
| Mouse Expression Set 430 | 1452652_PM_at | - | - |
| Mouse Expression Set 430 | 1428074_PM_at | - | - |
| Mouse Expression Set 430 | 1452652_at | - | - |
| Mouse Expression Set 430 | 1437750_PM_at | - | - |
| Mouse Expression Set 430 | 1437750_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437750_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437750_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428074_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452652_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428074_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452652_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428074_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428074_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437750_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437750_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452652_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452652_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 521080 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105270056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105270056 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160486 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2487322 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160486 | - | - |
| mouse-6 v1.1 (Illumina) | scl35175.1.186_300-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_50428573-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105270056 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105270056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160486 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160486 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_50428573-S | - | - |

Tmem158
Genomic Map