Assay Details
| Gene Symbol: Acvr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007397.2 | NP_031423.1 | 2 - 3 | 304 | 56 | |
| GenBank mRNA | AK137432.1 | - | 1 - 2 | 126 | 56 | |
| BC106189.1 | - | 2 - 3 | 361 | 30240596 | 56 | |
| M84120.1 | - | 2 - 3 | 302 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr2b
Genomic Map