Assay Details
| Gene Symbol: Acvr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007397.2 | NP_031423.1 | 8 - 9 | 1332 | 55 | |
| GenBank mRNA | AK137432.1 | - | 8 - 9 | 1084 | 55 | |
| BC106189.1 | - | 9 - 10 | 1317 | 30240596 | 55 | |
| M84120.1 | - | 8 - 9 | 1330 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr2b
Genomic Map