Assay Details
| Gene Symbol: Acvr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137432.1 | - | 3 - 4 | 386 | 86 | |
| BC106189.1 | - | 4 - 5 | 620 | 30240596 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acvr2b
Genomic Map