Assay Details
| Gene Symbol: Oxsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133985.2 | NP_598746.2 | 11 - 12 | 1424 | 73 | |
| NR_045693.1 | 12 - 13 | 1553 | 73 | |||
| GenBank mRNA | AK031790.1 | - | 10 - 11 | 921 | 73 | |
| AK075837.1 | - | 10 - 11 | 932 | 73 | ||
| AK129291.1 | - | 11 - 12 | 1419 | 73 | ||
| BC036148.1 | - | 2 - 3 | 174 | 73 | ||
| BC046453.1 | - | 12 - 13 | 1448 | 73 | ||
| BC060645.1 | - | 11 - 12 | 1407 | 6852021 | 73 | |
More Information
Set Membership: |
|

Oxsr1
Genomic Map