Assay Details
| Gene Symbol: Oxsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133985.2 | NP_598746.2 | 12 - 13 | 1459 | 115 | |
| NR_045693.1 | 13 - 14 | 1588 | 115 | |||
| GenBank mRNA | AK031790.1 | - | 11 - 12 | 956 | 115 | |
| AK075837.1 | - | 11 - 12 | 967 | 115 | ||
| AK129291.1 | - | 12 - 13 | 1454 | 115 | ||
| BC036148.1 | - | 3 - 4 | 209 | 115 | ||
| BC046453.1 | - | 13 - 14 | 1483 | 115 | ||
| BC060645.1 | - | 12 - 13 | 1442 | 6852021 | 115 | |
More Information
Set Membership: |
|

Oxsr1
Genomic Map