Assay Details
| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009371.3 | NP_033397.3 | 4 - 5 | 856 | 142 | |
| NM_029575.3 | NP_083851.3 | 3 - 4 | 781 | 142 | ||
| GenBank mRNA | AF406755.1 | - | 3 - 4 | 455 | 142 | |
| AK043619.1 | - | 4 - 5 | 940 | 142 | ||
| AK046561.1 | - | 4 - 5 | 782 | 142 | ||
| AK053793.1 | - | 4 - 5 | 857 | 142 | ||
| AK148167.1 | - | 3 - 4 | 781 | 142 | ||
| AK159327.1 | - | 4 - 5 | 836 | 142 | ||
| BC052629.1 | - | 758 | 142 | |||
| S69114.1 | - | 3 - 4 | 492 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbr2
Genomic Map