Assay Details
| Gene Symbol: 5033428I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017203.1 | - | 2 - 3 | 314 | 59 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60455 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01744590 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06522380 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01744607 | - | - |
| Whole Mouse Genome G4122A | A_52_P48557 | AK017203.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105205 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01744590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07407486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06913388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07359367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06913398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07094713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07407491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06840539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07407468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06694808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07166095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06840551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07374423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06913395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06990973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07374430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07009580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25890748 | - | - |

5033428I22Rik
Genomic Map