Assay Details
| Gene Symbol: Fblim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163256.1 | NP_001156728.1 | 7 - 8 | 1034 | 76 | |
| NM_133754.5 | NP_598515.3 | 7 - 8 | 1089 | 76 | ||
| GenBank mRNA | AK010660.1 | - | 7 - 8 | 1098 | 76 | |
| AK154680.1 | - | 7 - 8 | 1070 | 76 | ||
| AK163587.1 | - | 7 - 8 | 1075 | 76 | ||
| AK170151.1 | - | 7 - 8 | 1089 | 76 | ||
| AK170440.1 | - | 7 - 8 | 1039 | 76 | ||
| BC004777.1 | - | 6 - 7 | 964 | 3257486 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fblim1
Genomic Map