Assay Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145595.2 | NP_663570.2 | 2 - 3 | 380 | 131 | |
| GenBank mRNA | AK043313.1 | - | 2 - 3 | 205 | 131 | |
| AK150763.1 | - | 2 - 3 | 381 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbr4
Genomic Map