Assay Details
| Gene Symbol: Ndufa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023312.2 | NP_075801.1 | 3 - 4 | 946 | 59 | |
| GenBank mRNA | AF286698.1 | - | 2 - 3 | 181 | 59 | |
| AK002999.1 | - | 2 - 3 | 185 | 59 | ||
| AK003012.1 | - | 2 - 3 | 199 | 59 | ||
| AK010517.1 | - | 2 - 3 | 185 | 59 | ||
| AK012422.1 | - | 2 - 3 | 185 | 59 | ||
| BC034899.1 | - | 3 - 4 | 946 | 1447015 | 59 | |
| BC037149.1 | - | 2 - 3 | 189 | 5149812 | 59 | |
| BC055435.1 | - | 2 - 3 | 195 | 4219489 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufa13
Genomic Map