Assay Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253354.1 | NP_001240283.1 | 2 - 3 | 113 | 69 | |
| GenBank mRNA | BC046771.1 | - | 2 - 3 | 103 | 5703628 | 69 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC046771.1, NM_001253354.1 transcripts.
Set Membership: |
|

Fam214b
Genomic Map