Assay Details
| Gene Symbol: Slc6a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040692.3 | NP_001035782.1 | 5 - 6 | 821 | 76 | |
| NM_001136087.2 | NP_001129559.1 | 5 - 6 | 821 | 76 | ||
| NM_001168643.1 | NP_001162114.1 | 5 - 6 | 821 | 76 | ||
| NM_001168644.1 | NP_001162115.1 | 5 - 6 | 821 | 76 | ||
| NM_001168645.1 | NP_001162116.1 | 5 - 6 | 821 | 76 | ||
| NM_001168646.1 | NP_001162117.1 | 5 - 6 | 821 | 76 | ||
| GenBank mRNA | AF075262.1 | - | 5 - 6 | 798 | 76 | |
| AF075263.1 | - | 5 - 6 | 798 | 76 | ||
| AF075264.1 | - | 5 - 6 | 798 | 76 | ||
| AF075265.1 | - | 5 - 6 | 798 | 76 | ||
| AF075266.1 | - | 5 - 6 | 798 | 76 | ||
| AF075267.1 | - | 5 - 6 | 798 | 76 | ||
| AK085258.1 | - | 5 - 6 | 818 | 76 | ||
| AK085473.1 | - | 5 - 6 | 819 | 76 | ||
| AK128907.1 | - | 5 - 6 | 818 | 76 | ||
| BC010748.1 | - | 5 - 6 | 797 | 4235659 | 76 | |
More Information
Set Membership: |
|

Slc6a18
Genomic Map