Assay Details
| Gene Symbol: Slc6a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136087.2 | NP_001129559.1 | 8 - 9 | 1221 | 60 | |
| NM_001168645.1 | NP_001162116.1 | 8 - 9 | 1221 | 60 | ||
| GenBank mRNA | AF075264.1 | - | 8 - 9 | 1198 | 60 | |
| AF075266.1 | - | 8 - 9 | 1198 | 60 | ||
| AK085473.1 | - | 8 - 9 | 1219 | 60 | ||
| AK128907.1 | - | 8 - 9 | 1218 | 60 | ||
| BC010748.1 | - | 8 - 9 | 1197 | 4235659 | 60 | |
More Information
Set Membership: |
|

Slc6a18
Genomic Map