Assay Details
| Gene Symbol: Slc6a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136087.2 | NP_001129559.1 | 9 - 10 | 1309 | 71 | |
| NM_001168645.1 | NP_001162116.1 | 9 - 10 | 1309 | 71 | ||
| GenBank mRNA | AF075264.1 | - | 9 - 10 | 1286 | 71 | |
| AF075266.1 | - | 9 - 10 | 1286 | 71 | ||
| AK085473.1 | - | 9 - 10 | 1307 | 71 | ||
| AK128907.1 | - | 9 - 10 | 1306 | 71 | ||
| BC010748.1 | - | 9 - 10 | 1285 | 4235659 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a18
Genomic Map