Assay Details
| Gene Symbol: Slc6a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040692.3 | NP_001035782.1 | 10 - 11 | 1584 | 69 | |
| NM_001136087.2 | NP_001129559.1 | 10 - 11 | 1470 | 69 | ||
| NM_001168646.1 | NP_001162117.1 | 10 - 11 | 1554 | 69 | ||
| GenBank mRNA | AF075262.1 | - | 10 - 11 | 1561 | 69 | |
| AF075263.1 | - | 10 - 11 | 1531 | 69 | ||
| AF075264.1 | - | 10 - 11 | 1447 | 69 | ||
| AF075267.1 | - | 10 - 11 | 1500 | 69 | ||
| AK085473.1 | - | 10 - 11 | 1468 | 69 | ||
| AK128907.1 | - | 10 - 11 | 1467 | 69 | ||
| BC010748.1 | - | 10 - 11 | 1446 | 4235659 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a18
Genomic Map