Assay Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011977.3 | NP_036107.1 | 2 - 3 | 203 | 117 | |
| GenBank mRNA | BC028937.1 | - | 2 - 3 | 203 | 5362790 | 117 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC028937.1, NM_011977.3 transcripts.
Set Membership: |
|

Slc27a1
Genomic Map