Assay Details
| Gene Symbol: Gpr143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010951.3 | NP_035081.3 | 6 - 7 | 887 | 108 | |
| GenBank mRNA | AK185813.1 | - | 3 - 4 | 320 | 108 | |
| BC119384.1 | - | 6 - 7 | 803 | 8734133 | 108 | |
| BC137813.1 | - | 6 - 7 | 803 | 8860832 | 108 | |
| U63918.1 | - | 6 - 7 | 822 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr143
Genomic Map