Assay Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030035.1 | NP_084311.1 | 10 - 11 | 1595 | 105 | |
| GenBank mRNA | BC145221.1 | - | 10 - 11 | 1554 | 9053772 | 105 |
| BC150731.1 | - | 10 - 11 | 1572 | 9087642 | 105 | |
More Information
Set Membership: |
|

Golgb1
Genomic Map