Assay Details
| Gene Symbol: Faf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178397.3 | NP_848484.2 | 2 - 3 | 185 | 76 | |
| GenBank mRNA | AK051724.1 | - | 2 - 3 | 152 | 76 | |
| AK122394.1 | - | 2 - 3 | 81 | 76 | ||
| AK148529.1 | - | 2 - 3 | 146 | 76 | ||
| AK151617.1 | - | 2 - 3 | 147 | 76 | ||
| AK170111.1 | - | 2 - 3 | 169 | 76 | ||
| AK171922.1 | - | 2 - 3 | 169 | 76 | ||
| BC046817.1 | - | 2 - 3 | 150 | 5702561 | 76 | |
| BC048767.1 | - | 3 - 4 | 209 | 76 | ||
| BY139990.1 | - | 2 - 3 | 185 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faf2
Genomic Map