Assay Details
| Gene Symbol: Faf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178397.3 | NP_848484.2 | 3 - 4 | 325 | 102 | |
| GenBank mRNA | AK122394.1 | - | 3 - 4 | 221 | 102 | |
| AK148529.1 | - | 3 - 4 | 286 | 102 | ||
| AK170111.1 | - | 3 - 4 | 309 | 102 | ||
| BC046817.1 | - | 3 - 4 | 287 | 5702561 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Faf2
Genomic Map