Assay Details
| Gene Symbol: Faf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178397.3 | NP_848484.2 | 6 - 7 | 631 | 79 | |
| GenBank mRNA | AK122394.1 | - | 6 - 7 | 527 | 79 | |
| AK148529.1 | - | 6 - 7 | 592 | 79 | ||
| AK151617.1 | - | 6 - 7 | 536 | 79 | ||
| AK170111.1 | - | 6 - 7 | 615 | 79 | ||
| BC046817.1 | - | 6 - 7 | 593 | 5702561 | 79 | |
| BC048767.1 | - | 5 - 6 | 439 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Faf2
Genomic Map