Assay Details
| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083902.1 | NP_001077371.1 | 7 - 8 | 940 | 63 | |
| GenBank mRNA | AK165080.1 | - | 3 - 4 | 563 | 63 | |
| BC030926.1 | - | 7 - 8 | 940 | 4910321 | 63 | |
More Information
Set Membership: |
|

Slc12a8
Genomic Map