Assay Details
| Gene Symbol: Gne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190414.1 | NP_001177343.1 | 7 - 8 | 1631 | 127 | |
| NM_015828.3 | NP_056643.3 | 7 - 8 | 1449 | 127 | ||
| GenBank mRNA | AJ132236.1 | - | 6 - 7 | 1277 | 127 | |
| AK010070.1 | - | 4 - 5 | 597 | 127 | ||
| AK136573.1 | - | 7 - 8 | 1450 | 127 | ||
| AK170707.1 | - | 7 - 8 | 1632 | 127 | ||
| AM697710.1 | - | 7 - 8 | 1370 | 127 | ||
| BC015277.1 | - | 7 - 8 | 1437 | 4216299 | 127 | |
| BC051254.1 | - | 7 - 8 | 1437 | 6478532 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gne
Genomic Map