Assay Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159633.1 | NP_001153105.1 | 11 - 12 | 1678 | 73 | |
| NM_133891.3 | NP_598652.3 | 11 - 12 | 1678 | 73 | ||
| GenBank mRNA | AK077951.1 | - | 11 - 12 | 1681 | 73 | |
| AY249865.1 | - | 11 - 12 | 1434 | 73 | ||
| AY249866.1 | - | 11 - 12 | 1407 | 73 | ||
| BC025941.1 | - | 2 - 3 | 240 | 73 | ||
| BC113167.1 | - | 11 - 12 | 1434 | 40059854 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025941.1 transcripts.
Set Membership: |
|

Slc44a1
Genomic Map