Assay Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159633.1 | NP_001153105.1 | 3 - 4 | 534 | 81 | |
| NM_133891.3 | NP_598652.3 | 3 - 4 | 534 | 81 | ||
| GenBank mRNA | AY249865.1 | - | 3 - 4 | 290 | 81 | |
| AY249866.1 | - | 3 - 4 | 263 | 81 | ||
| BC113167.1 | - | 3 - 4 | 290 | 40059854 | 81 | |
| BC113169.1 | - | 3 - 4 | 290 | 40059860 | 81 | |
| CF532681.1 | - | 2 - 3 | 165 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc44a1
Genomic Map