Assay Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159633.1 | NP_001153105.1 | 7 - 8 | 1029 | 76 | |
| NM_133891.3 | NP_598652.3 | 7 - 8 | 1029 | 76 | ||
| GenBank mRNA | AK077951.1 | - | 7 - 8 | 1032 | 76 | |
| AK200057.1 | - | 2 - 3 | 187 | 76 | ||
| AY249865.1 | - | 7 - 8 | 785 | 76 | ||
| AY249866.1 | - | 7 - 8 | 758 | 76 | ||
| BC113167.1 | - | 7 - 8 | 785 | 40059854 | 76 | |
| BC113169.1 | - | 7 - 8 | 785 | 40059860 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc44a1
Genomic Map