Assay Details
| Gene Symbol: Tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028053.2 | NP_082329.1 | 2 - 3 | 322 | 94 | |
| GenBank mRNA | AB261159.1 | - | 2 - 3 | 258 | 94 | |
| AK005479.1 | - | 2 - 3 | 305 | 94 | ||
| AK136131.1 | - | 1 - 2 | 72 | 94 | ||
| AK150377.1 | - | 2 - 3 | 300 | 94 | ||
| AK150892.1 | - | 2 - 3 | 300 | 94 | ||
| AK153075.1 | - | 2 - 3 | 287 | 94 | ||
| BC011072.1 | - | 2 - 3 | 300 | 3978742 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem38b
Genomic Map