| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400014074.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014074.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014074.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014074.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014074.mm.1 |
TC0400001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014079.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014079.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014079.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014079.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014079.mm.1 |
TC0400001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014075.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014075.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014075.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014075.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014075.mm.1 |
TC0400001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400014077.mm.1 |
NM_001190978.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014077.mm.1 |
NM_001190978.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014077.mm.1 |
NM_010623.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014077.mm.1 |
NM_010623.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400014077.mm.1 |
TC0400001721.mm.1 |
MTA TC |