Assay Details
| Gene Symbol: Kif17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190978.1 | NP_001177907.1 | 10 - 11 | 2284 | 95 | |
| NM_010623.4 | NP_034753.1 | 12 - 13 | 2995 | 95 | ||
| GenBank mRNA | AB008867.1 | - | 12 - 13 | 2993 | 95 | |
| AK077868.1 | - | 10 - 11 | 2065 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif17
Genomic Map