Assay Details
| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144904.2 | NP_659153.2 | 4 - 5 | 493 | 153 | |
| NM_178164.3 | NP_835458.1 | 3 - 4 | 468 | 153 | ||
| GenBank mRNA | AK033732.1 | - | 3 - 4 | 393 | 153 | |
| AK088560.1 | - | 3 - 4 | 400 | 153 | ||
| BC006638.1 | - | 4 - 5 | 403 | 3969488 | 153 | |
| BC057641.1 | - | 3 - 4 | 373 | 4936557 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp3
Genomic Map