Assay Details
| Gene Symbol: Hsdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301367.1 | NP_001288296.1 | 5 - 6 | 691 | 123 | |
| NM_024255.3 | NP_077217.2 | 4 - 5 | 593 | 123 | ||
| GenBank mRNA | AJ293845.1 | - | 5 - 6 | 614 | 124 | |
| AK011910.1 | - | 4 - 5 | 551 | 123 | ||
| AK085899.1 | - | 4 - 5 | 562 | 123 | ||
| AK136773.1 | - | 4 - 5 | 560 | 123 | ||
| AK144948.1 | - | 4 - 5 | 509 | 123 | ||
| AK145230.1 | - | 4 - 5 | 453 | 123 | ||
| AY725196.1 | - | 4 - 5 | 450 | 123 | ||
| BC002211.1 | - | 4 - 5 | 466 | 3490303 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ293845.1, NM_001301367.1 transcripts.
Set Membership: |
|

Hsdl2
Genomic Map