Assay Details
| Gene Symbol: Cldnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171826.3 | NP_741968.1 | 1 - 2 | 176 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_171826.3 transcripts.
Set Membership: |
|

Cldnd1
Genomic Map