Assay Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019436.3 | NP_062309.2 | 3 - 4 | 363 | 67 | |
| GenBank mRNA | AJ236881.1 | - | 3 - 4 | 339 | 67 | |
| AK079882.1 | - | 3 - 4 | 365 | 67 | ||
| BC064055.1 | - | 3 - 4 | 376 | 576098 | 67 | |
More Information
Set Membership: |
|

Sit1
Genomic Map