Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 9 - 10 | 680 | 100 | |
| GenBank mRNA | AK029992.1 | - | 10 - 11 | 910 | 100 | |
| AK076708.1 | - | 9 - 10 | 681 | 100 | ||
| AK078567.1 | - | 9 - 10 | 649 | 100 | ||
| EF377342.1 | - | 9 - 10 | 669 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b5
Genomic Map