| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400002691.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002691.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002691.mm.1 |
TC0400000425.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400002692.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002692.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002692.mm.1 |
TC0400000425.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400002688.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002688.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002688.mm.1 |
TC0400000425.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400002689.mm.1 |
NM_172690.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002689.mm.1 |
NM_172690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400002689.mm.1 |
TC0400000425.mm.1 |
MTA TC |