Assay Details
| Gene Symbol: N28178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172690.2 | NP_766278.1 | 7 - 8 | 1037 | 84 | |
| GenBank mRNA | AK038528.1 | - | 6 - 7 | 891 | 84 | |
| AK046283.1 | - | 7 - 8 | 1028 | 84 | ||
| AK122428.1 | - | 5 - 6 | 960 | 84 | ||
| BC057092.1 | - | 7 - 8 | 1037 | 5703966 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

N28178
Genomic Map