Assay Details
| Gene Symbol: Mcur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081059.3 | NP_001074528.1 | 1 - 2 | 426 | 109 | |
| GenBank mRNA | AK018089.1 | - | 1 - 2 | 397 | 109 | |
| AK041849.1 | - | 1 - 2 | 427 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcur1
Genomic Map