Assay Details
| Gene Symbol: Mcur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081059.3 | NP_001074528.1 | 3 - 4 | 651 | 147 | |
| GenBank mRNA | AK018089.1 | - | 3 - 4 | 622 | 147 | |
| AK041849.1 | - | 3 - 4 | 652 | 147 | ||
| BC113195.1 | - | 4 - 5 | 389 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC113195.1 transcripts.
Set Membership: |
|

Mcur1
Genomic Map