Assay Details
| Gene Symbol: Mcur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081059.3 | NP_001074528.1 | 4 - 5 | 741 | 148 | |
| GenBank mRNA | AK018089.1 | - | 4 - 5 | 712 | 148 | |
| AK041849.1 | - | 4 - 5 | 742 | 148 | ||
| BC113195.1 | - | 5 - 6 | 479 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcur1
Genomic Map