Assay Details
| Gene Symbol: Mcur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081059.3 | NP_001074528.1 | 6 - 7 | 867 | 95 | |
| GenBank mRNA | AK018089.1 | - | 6 - 7 | 838 | 95 | |
| BC113195.1 | - | 7 - 8 | 605 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcur1
Genomic Map