Assay Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181728.2 | NP_859417.2 | 1 - 2 | 71 | 75 | |
| GenBank mRNA | AK016257.1 | - | 1 - 2 | 72 | 75 | |
| DQ519368.1 | - | 1 - 2 | 70 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the DQ519368.1, NM_181728.2 transcripts.
Set Membership: |
|

Art3
Genomic Map