Assay Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271397.1 | NP_001258326.1 | 3 - 4 | 815 | 134 | |
| NR_073167.1 | 3 - 4 | 996 | 134 | |||
| NR_073168.1 | 3 - 4 | 811 | 134 | |||
| GenBank mRNA | AK014994.1 | - | 3 - 4 | 816 | 134 | |
| AK029961.1 | - | 3 - 4 | 528 | 134 | ||
| AK147172.1 | - | 3 - 4 | 551 | 134 | ||
| BC049166.1 | - | 3 - 4 | 406 | 134 | ||
| BC058699.1 | - | 3 - 4 | 971 | 134 | ||
| BC125575.1 | - | 3 - 4 | 541 | 40130090 | 134 | |
| BC125579.1 | - | 3 - 4 | 541 | 40130094 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nol8
Genomic Map